Order Entry
Puerto Rico
ContactUsLinkComponent
Pierce™ DyLight™ Fluor-Free Acids, Antibody Labelling Dyes, Thermo Scientific
Pierce™ DyLight™ Fluor-Free Acids, Antibody Labelling Dyes, Thermo Scientific
Catalog # PI62280
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ DyLight™ Fluor-Free Acids, Antibody Labelling Dyes, Thermo Scientific
Catalog # PI62280
Supplier:  Invitrogen
Supplier Number:  62280
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    DyLight™ 488 free acid
  • Size:
    1 mg
  • Storage Conditions:
    Store at < 4˚C.
  • Cat. No.:
    PI62280
  • Supplier no.:
    62280

Specifications

About this item

DyLight™ fluor-free acids are non-activated forms of selected DyLight™ fluorescent dyes for use as controls and calibration factors in fluorescence imaging applications

These non-activated fluorescent dyes contain the native carboxyl group of the standard dye molecule.

Caution: For Research Use Only. Not for use in diagnostic procedures.