Order Entry
Puerto Rico
ContactUsLinkComponent
 
Pierce™ Thiophilic Adsorption Kit, Thermo Scientific
Catalog # PI44916
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ Thiophilic Adsorption Kit, Thermo Scientific
Catalog # PI44916
Supplier:  Invitrogen
Supplier Number:  44916
CAS Number:  

Specifications

  • Description:
    Thiophilic Adsorption Kit, 4 columns, sufficient for purifying up to 60 mg Immunoglobulin per column
  • Size:
    4 columns
  • Storage Temperature:
    Store at 4 ˚C.
  • Description:
    T-Gel purification kit
  • Cat. No.:
    PI44916
  • Supplier no.:
    44916

Specifications

About this item

Thermo Scientific Pierce Thiophilic Adsorbent is a protein-free immunosorbent resin for gentle preferential binding and purification of immunoglobulins from a variety of sources including mouse, rat, rabbit, goat and human.

  • High capacity - gently binds and purifies
  • Broad antibody specificity
  • Simple and fast - quickly fractionate and purify the immunoglobulin component
  • Binding condition is non denaturing and the elution buffer yields concentrated, salt-free immunoglobulin

This Pierce Thiophilic Adsorption Kit is a purification kit containing columns pre-packed with the resin and ready-to-use buffers.

Pierce Thiophilic Adsorbent is 6% beaded agarose that has been modified to contain simple sulfone-thioether groups. The result is an affinity resin that has a high binding capacity and a broad specificity for many species of immunoglobulins when incubated in 0.5M potassium sulfate. In effect, thiophilic adsorption is a resin-based variation of ammonium sulfate precipitation for quick immunoglobulin purification from crude serum samples. Unlike antibody fractionation by ammonium sulfate precipitation, purification with thiophilic adsorbent yields concentrated, essentially salt-free, highly purified immunoglobulins at near-neutral pH. Thus, this simple one-step method eliminates the need for additional treatment of the sample for storage or for subsequent conjugation reactions. The gentle binding and elution conditions ensure a high protein recovery with excellent preservation of antibody activity.

Thiophilic adsorption is based on the ability of some proteins, particularly immunoglobulins, to bind to an immobilized ligand that contains a sulfone group in close proximity to a thioether. Salts that interact with water molecules, such as potassium and ammonium sulfate, promote this binding by driving the protein into close proximity with the ligand.

Caution: For Research Use Only. Not for use in diagnostic procedures.