Order Entry
Puerto Rico
ContactUsLinkComponent
Pierce™ CarboxyLink™ Immobilization Kit, 2 ml, Thermo Scientific
Pierce™ CarboxyLink™ Immobilization Kit, 2 ml, Thermo Scientific
  PI44899
 :  
undefined
Pierce™ CarboxyLink™ Immobilization Kit, 2 ml, Thermo Scientific
  PI44899
 :  44899
 :  

 

  • Description:
    CarboxyLink™ Immobilization Kit
  • Size:
    5 columns
  • Cat. No.:
    PI44899
  • Supplier no.:
    44899
  • Sufficient for:
    Immobilizng 1 - 2 mg of Peptide per Column

 

 

The Thermo Scientific CarboxyLink Immobilization Kit is for covalent immobilization of peptides or other carboxyl-containing (—COOH) molecules to a porous, beaded resin for use in affinity purification procedures.

  • Ideal for unmodified peptides, immobilizes peptides with high capacity and various orientations without steric hindrance
  • DADPA-activated crosslinked 4% beaded agarose acting as a CarboxyLink™ coupling resin
  • With stable linkage, flexible and gentle coupling conditions
  • Sufficient for Immobilizing 1 to 2 mg of peptide per column

CarboxyLink Resin is crosslinked beaded agarose that has been activated with diamino-dipropylamine (DADPA) to contain long spacer arms, each with a primary amine at the end

Because EDC crosslinks carboxylates to primary amines, peptides and other molecules that contain both of these functional groups will be variously polymerized as well as immobilized to the CarboxyLink Resin. For peptides whose only carboxyl and amino groups are at the termini, coupling with the CarboxyLink Kit will result in end-to-end chains of peptide that are linked by their C-terminal ends to the support. Generally, this structure provides for high capacity and low steric hindrance for use in affinity purification. However, in the case of peptides containing multiple primary amines (lysine) or carboxylates (glutamic and aspartic acid) in the middle of their sequence, the CarboxyLink Coupling Method may cause key binding sites to be blocked, resulting in high coupling capacity but poor binding capacity for affinity purification.

When incubated with the resin and the carbodiimide crosslinker EDC (included in the CarboxyLink Immobilization Kit), carboxyl-containing molecules become permanently attached to the support by stable amide bonds. Once a molecule is coupled, the prepared affinity column can be used multiple times in typical protein affinity purification procedures. CarboxyLink Coupling Resins can also be used to immobilize other kinds of molecules using alternative amine-reactive crosslinking chemistries.

 : CarboxyLink Columns, 2 mL, 5 columns, EDC Crosslinker, 5 x 60 mg, MES Buffered Saline Pack (makes 500 mL), 1 pack, Wash Solution, 120 mL, Column Accessories
 : For Research Use Only. Not for use in diagnostic procedures. Store EDC desiccated at -20˚C. Store other components at 4˚C.