Pierce™ Fast Wash Buffer, 10X, Thermo Scientific
Catalog # PI37577
Supplier: Invitrogen
CAS Number:
Specifications
- Description:Pierce™ Fast Wash Buffer, 10X, Washing Buffer
- Size:1 L
- Cat. No.:PI37577
- Supplier no.:37577
Specifications
About this item
Pierce Fast Wash Buffer, 10X, for use with Pierce Fast Blocking Buffer.
- Simple
- Low background
Pierce Fast Blocking Buffer streamlines Western blot protocols by providing consistent, high-quality blocking for nitrocellulose or PVDF membranes in only 5 minutes. Blocking and probing steps with Fast Blocking Buffer prevents nonspecific binding of detection antibodies during probing steps but allows specific detection to occur.
Caution: For Research Use Only. Not for use in diagnostic procedures. Store at 4˚C.