Order Entry
Puerto Rico
ContactUsLinkComponent
Pierce™ ONPG (o-nitrophenyl-β-D-galactopyranoside), Thermo Scientific
Pierce™ ONPG (o-nitrophenyl-β-D-galactopyranoside), Thermo Scientific
Catalog # PI34055
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ ONPG (o-nitrophenyl-β-D-galactopyranoside), Thermo Scientific
Catalog # PI34055
Supplier:  Invitrogen
Supplier Number:  34055
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ONPG, powder
  • Size:
    5 g
  • Storage Conditions:
    Upon receipt store at -20˚C protected from moisture and light
  • Cat. No.:
    PI34055
  • Supplier no.:
    34055

Specifications

About this item

Thermo Scientific Pierce ONPG is the preferred colorimetric substrate for ELISA applications involving beta-galactosidase (b-Gal) as the reporter enzyme.

  • Product formed is completely soluble
  • High extinction coefficient at 405nm
  • Substrate yields a yellow product that is easily detectable in the visual range after stopping the reaction with 1M sodium carbonate

When using beta-Galactosidase as the label for proteins in ELISA studies, a wide variety of substrates are available, including: o-Nitrophenyl-beta-D-galactopyranosidase (ONPG), Naphthol-AS-Bl-beta-D-galactopyranosidase (Nap-Gal), and 4-Methyl-umbelliferyl-beta-D-galactopyranosidase (MUm-Gal). However, it is important to choose a substrate with adequate solubility, that uses readily available equipment, and that gives a significant reading over the background. ONPG is a superior beta-Galactosidase substrate option. The product formed is completely soluble and has a high extinction coefficient at 405nm. The substrate yields a yellow product that is easily detectable in the visual range after stopping the reaction with 1M Sodium Carbonate.

Caution: For Research Use Only. Not for use in diagnostic procedures.