
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate) substrate ≥99%, powder, Pierce™
Catalog # PI34040
Supplier: Invitrogen
CAS Number:
Specifications
- Size:1 g
- Cat. No.:PI34040
- Supplier no.:34040
Specifications
Specification Test Results
- VISUAL:White to off-white crystalline solid, free of foreign matter.
- WATER CONTENT:(KF) ≤ 2%
- MOLAR ABSORPTIVITY:≥ 5700 at ~292 nm in 10mM Diethanolamine, pH 10.3 at 25 °C
- PURITY:(HPLC) ≥ 99%
- INORGANIC PHOSPHATE:≤ 0.1%
Specification Test Results
About this item
:
:433.6214 g/mol
EINECS:229-506-1
:GHS07
Storage Temperature:Freezer
CAS Number:6578-06-9
Formula:C₁₅H₁₅BrClN₂O₄P
MDL Number:MFCD00040636