Pierce™ HRP Substrates for ELISA, OPD (o-phenylenediamine dihydrochloride), Thermo Scientific
Thermo Scientific Pierce OPD is a water-soluble substrate for horseradish peroxidase (HRP) that produces a yellow-orange product detectable at 492nm by ELISA plate readers.
- Water soluble product when reacted with peroxidase
- Absorbance maximum of 492nm
- Upon receipt store OPD powder at 4˚C and OPD tablets at -20˚C
- Product is shipped at ambient temperature
OPD yields a water soluble yellow-orange product when reacted with peroxidase with an absorbance maximum of 492. OPD can easily be dissolved in a substrate buffer such as Stable Peroxide Substrate Buffer (Part No. 34062).
: For Research Use Only. Not for use in diagnostic procedures.
- Form:powder