Order Entry
Puerto Rico
ContactUsLinkComponent
Pierce™ CN/DAB Substrate Kit, Thermo Scientific
Pierce™ CN/DAB Substrate Kit, Thermo Scientific
Catalog # PI34000
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ CN/DAB Substrate Kit, Thermo Scientific
Catalog # PI34000
Supplier:  Invitrogen
Supplier Number:  34000
CAS Number:  

Specifications

  • Description:
    CN/DAB Substrate Kit, sufficient for 25 uses at 10 mL each
  • Size:
    275 mL
  • Storage Conditions:

    4˚C
  • Cat. No.:
    PI34000
  • Supplier no.:
    34000

Specifications

About this item

Thermo Scientific Pierce CN/DAB Substrate Kit includes substrate and peroxide solutions for combined chloronaphthol- and diaminobenzidine-based detection of horseradish peroxidase (HRP) activity in Western blot and tissue staining methods.

  • HRP substrate—for detection of horseradish peroxidase activity on solid media via the combined action of CN (4-chloro-1-naphthol) and DAB (3,3'-diaminobenzidine tetrahydrochloride)
  • Chromogenic—no special equipment needed for visualization; yields intense black bands that are easy to photograph
  • Easy-to-use kit—substrate stock solution and stable peroxide substrate buffer mean that there's no dry powders to measure and dissolve
  • Stable—store refrigerated at 4˚C for at least one year

This kit combines two popular chromogenic peroxidase substrate compounds (4-CN and DAB) in a single, stable, 10X stock solution. This CN/DAB solution produces an intense dark black precipitate at sites of bound HRP-conjugated antibodies on probed blots and tissue samples. The Pierce CN/DAB Substrate Kit can be used in Western immunoblotting techniques as well as in dot blotting assays. The kit includes the 10X solution of CN/DAB and accompanying 1X Stable Peroxide Substrate Buffer.

Delivery: CN/DAB Solution (10X) and Stable Peroxide Substrate Solution (1X)
Caution: For Research Use Only. Not for use in diagnostic procedures.