Order Entry
United States
ContactUsLinkComponent
 
Dulbecco's Modified Eagle's Limiting Medium (DMEM-LM), Thermo Scientific
  PI30030
 :  
Dulbecco's Modified Eagle's Limiting Medium (DMEM-LM), Thermo Scientific
  PI30030
 :  30030
 :  

 

  • Description:
    Dulbecco'S Modified Eagle'S
  • Size:
    500 ml
  • Environmentally Preferable:
  • Storage Temperature:
    4 °C
  • Cat. No.:
    PI30030
  • Supplier no.:
    30030

 

 

Dulbecco's Modified Eagle Medium (DMEM) is for supporting and maintening a broad spectrum of mamamalian cell types.

  • Efficient recovery
  • Increased specificity compared to traditional methods
  • Easy to use

Formulation: Sterile filtered DMEM (-)L-leucine, (-)L-methionine with 4.5 g/L glucose, 4.0 mM L-glutamine, sodium pyruvate and phenol red.

 : For Research Use Only. Not for use in diagnostic procedures.