ContactUsLinkComponent
Pierce™ 10X TBE Buffer, Thermo Scientific
Pierce™ 10X TBE Buffer, Thermo Scientific
Catalog # PI28355
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ 10X TBE Buffer, Thermo Scientific
Catalog # PI28355
Supplier:  Invitrogen
Supplier Number:  28355
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    TBE Buffer Solution 10X Concentrate (TRIS-Borate-EDTA Buffer)
  • Size:
    1 L
  • Environmentally Preferable:
  • Cat. No.:
    PI28355
  • Supplier no.:
    28355
Specifications
About this item
Thermo Scientific Pierce 10X TBE Buffer is a space-saving stock solution that is ideal for casting and preparing Tris-Borate-EDTA (pH 8.3) running buffer used for agarose and polyacrylamide gel electrophoresis of nucleic acids.
  • TBE buffer—diluted 10-fold in water, the solution yields 0.089M Tris, 0.089M borate, 2 mM EDTA, pH 8.2 to 8.4
  • Easy to use—no packets to open and no powder to dissolve
  • Increased accuracy—eliminates the possibility of powder remaining in a packet
  • Saves time—10X concentration eliminates time spent waiting for powder to dissolve
  • Saves space—storage as concentrated stock minimizes bench space needed for solutions
Pierce Concentrated Buffers are ready to use without having to weigh and dissolve dry ingredients or to adjust the pH with concentrated acid or base. Simply dilute the stock solution with pure water and proceed with your experiment. The 10X TBE Buffer (Tris-borate-EDTA) makes 0.089M Tris, 0.089M borate, 2 mM EDTA, pH 8.2 to 8.4, when diluted to 1X with water.
Ordering information: Store at room temperature.
Caution: For Research Use Only. Not for use in diagnostic procedures.