Order Entry
Puerto Rico
ContactUsLinkComponent
Pierce™ PITC, Edmans Reagent, Thermo Scientific
Pierce™ PITC, Edmans Reagent, Thermo Scientific
Catalog # PI26922
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ PITC, Edmans Reagent, Thermo Scientific
Catalog # PI26922
Supplier:  Invitrogen
Supplier Number:  26922
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    PITC (Edman’s reagent)
  • Size:
    10 x 1 mL
  • Cat. No.:
    PI26922
  • Supplier no.:
    26922

Specifications

About this item

This Phenylisothiocyanate (PITC) is a high-purity reagent for pre-column quantitative derivatization of amino acids by reverse-phase HPLC.

  • For pre-column derivatization of amino acids for picomole detection levels in HPLC
  • Reacts with amine groups of amino acids in 5 to 10 minutes at room temperature to yield products that are detectable by UV absorbance (λmax = 254nm)
  • Produces phenylthiocarbamyl (PTC) derivatives that can be separated and quantified in 30 minutes using reverse-phase HPLC
  • Produces stable products with all amino acids, including proline

Phenylisothiocyanate (PITC), also known as Edman's Reagent, enables the sequential degradation of amino acids in a polypeptide chain, yielding primary structural information.

Reagent Properties:
Alternative names: PITC, Phenyl Isothiocyanate, Edman's Reagent
Chemical name: Isothiocyanatobenzene
Reactive toward: Primary amines (—NH2)
Chemical formula: C₇H₅NS
CAS number: 103-72-0
Molecular weight: 135.19
Form: Clear, colorless to pale yellow liquid
Storage: Upon receipt store at room temperature

Unlike Fmoc-chloride, PITC does not yield disubstituted tyrosine or histidine derivatives. PTC-amino acids demonstrate improved stability at pH 5-7.5 as well as increased stability at room temperature over o-phthalaldehyde (OPA)-amino acid adducts. Also, unlike OPA, PITC enables the direct analysis of secondary amino acids.

Caution: For Research Use Only. Not for use in diagnostic procedures.