Specifications
- Concentration:1 mg/ml
- Description:Peptide digest assay standard
- Size:1.5 mL
- Cat. No.:PI23295
- Supplier no.:23295
Specifications
About this item
Thermo Scientific™ Peptide Digest Assay Standard is a validated protein digest reference sample optimized for use in quantitative peptide assays to improve reproducibility and accuracy.Robust peptide assay standard—the only commercially available peptide digest standard that has been developed for use in a quantitative peptide assayValidated—validated for both the Pierce Quantitative Fluorometric Peptide Assay and the Pierce Quantitative Colorimetric Peptide Assay with comparable standard curves using both assaysStable—liquid formulation is stored at 4⁰C, eliminating variability due to freeze-thawsEasy-to-use—provided in a liquid format with a known peptide concentrationThe Peptide Digest Assay Standard was designed as reference standard for use with the Pierce Quantitative Fluorometric and Colorimetric Peptide Assays to augment the reproducibility and accuracy of peptide quantitation in a microplate-based assay.The reference protein has been digested with MS-grade trypsin to minimize missed cleavages
To help ensure consistent performance, digestion efficiency of the protein is monitored to help ensure lot-to-lot consistency, and quality is assessed by comparison to a reference standard. The Peptide Digest Assay Standard is provided in ready-to-use liquid format at 1 mg/mL, and is sufficient for at least ten standard curves (depending on dilution volumes).
Store in refrigerator (2–8˚C).