Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Size:500 assays
- Cat. No.:PI23290
- Supplier no.:23290
The Thermo Scientific™ Pierce™ Quantitative Fluorescent Peptide Assay is a sensitive, mix-and-read fluorescent microplate assay for the quantitative measurement of peptides and peptide mixtures.Sensitive—accurately detect as little as 5.0 µg/mL of single peptides or peptide mixturesRobust— assay performance rigorously tested using both peptides and peptide digest mixturesRobust peptide digest standard—kit includes a validated peptide digest standard for improved reproducibility of quantitationCompatible—works with many reagents, including those used in mass spectrometry sample preparationConvenient—easy mix-and-read format and stable fluorescent signal that may be read in as little as 5 minutes up to several hoursThe Pierce Quantitative Fluorescent Peptide Assay reagents include peptide assay buffer, fluorescent peptide labeling reagent, and a peptide digest assay standard for the quantitative measurement of peptide concentrations
In this assay, peptides are specifically labeled at the amino-terminus using an amine-reactive fluorescent reagent, and the fluorescently labeled peptides are detected using ex390/em475.Because of the labeling mechanism of the fluorescent assay reagent, this assay is suitable for the quantitative measurement of synthetic peptides as well as peptide digest mixtures. A high quality peptide digest reference standard is included for use in generating linear standard curves and as a calibration control for improved accuracy and reproducibility. This sensitive assay requires only 10 µL of sample, produces a linear response with increasing peptide concentrations (5–1000 µg/mL), and results in a stable final fluorescence that can be detected in as little as five minutes.Applications:Recommended for use with peptide mixtures and synthetic peptides with an unblocked amino-terminus (amino-terminus-labeled peptides, such as TMT-labeled peptides, are not compatible with this assay)Normalizing sample concentrations for quantitative downstream applicationsNormalizing sample loading volumes for LC, MS, and LC/MS applicationsMeasuring recovery after peptide clean-up procedures
Fluorometric Peptide Assay Buffer, 50 mL
Peptide Digest Assay Standard, 1.5 mL
Store in refrigerator (2–8˚C).