Order Entry
Puerto Rico
ContactUsLinkComponent
Pierce™ Quantitative Colorimetric Peptide Assay
Pierce™ Quantitative Colorimetric Peptide Assay
Catalog # PI23275
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ Quantitative Colorimetric Peptide Assay
Catalog # PI23275
Supplier:  Invitrogen
Supplier Number:  23275
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Pierce™ Quantitative colorimetric peptide assay
  • Size:
    500 assays
  • Cat. No.:
    PI23275
  • Supplier no.:
    23275

Specifications

About this item

Thermo Scientific™ Pierce™ Quantitative Colorimetric Peptide Assay is an easy-to-use colorimetric microplate assay designed specifically to improve the sensitivity and reproducibility of quantitation of peptide mixtures.

  • Sensitive – accurately detect as little as 25 µg/ml of peptide mixture
  • Robust – assay performance rigorously tested using peptide digest mixtures
  • Robust peptide digest standard – kit includes a validated peptide digest standard for improved reproducibility of quantitation
  • Compatible – works with many reagents, including mass spectrometry sample preparation reagents and TMT™-labeled peptides
  • Convenient – easy mix-and read-format and stable colorimetric signal that may be read in as little as 15 minutes

The Pierce Quantitative Colorimetric Peptide Assay provides modified BCA reagents for the reduction of Cu+2 to Cu+1 and a proprietary chelator that have been optimized for the quantitation of peptide mixtures. In this reaction, the copper is first reduced by the amide backbone of peptides under alkaline conditions (Biuret reaction). The proprietary chelator then couples with the reduced copper to form a bright red complex with absorbance of 480 nm. The signal produced from this reaction is 3-4 fold more sensitive than the Micro-BCA Protein Assay for peptide analysis.

The kit also contains a high quality peptide digest reference standard for use in generating linear standard curves and to serve as a calibration control. This colorimetric peptide assay requires a small amount of sample (20 µl) and has a working peptide concentration range of 25 to 1000 μg/ml. The increased sensitivity, low sample assay volume, and included reference standard are essential for accurate and robust measurement of peptide digest samples, especially for mass spectrometry applications.

Applications:
Recommended for peptide mixtures
Not recommended for synthetic peptides because the assay is affected by peptide amino acid content
Normalizing sample concentrations for quantitative downstream applications
Normalizing sample loading volumes for LC, MS, and LC/MS applications
Measuring recovery after peptide clean-up procedures

Storage: 2 to 8 °C

Ordering information: Kit contains Colorimettric Assay Reagent A, 50 ml; Colorimetric Assay Reagent B, 2×25 ml; Colorimetric Assay Reagent C, 2 ml; Peptide Digest Assay Standard, 1,5 ml.
Caution: For Research Use Only. Not for use in diagnostic procedures.