Boronic Acid Resin, Pierce™
Catalog # PI20244
Supplier: Invitrogen
CAS Number:
Specifications
- Size:10 ml
- Cat. No.:PI20244
- Supplier no.:20244
Specifications
Specification Test Results
- VISUAL:Off-white gel slurry, free of foreign matter
- ASSAY: % Recovery of AMP:95 - 105%
Specification Test Results
About this item
:
Storage Temperature:Refrigerator