Bovine Tpck Trypsin (from Pancreas)
Catalog # PI20233
Supplier: Invitrogen
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- CAS No.:9002-07-7
- Source:Pancreas
- Species:Bovine
- Size:50 mg
- Storage Conditions:Refrigerator
- Enzyme Name:Tpck Trypsin
- Cat. No.:PI20233
- Supplier no.:20233
Specifications
About this item
Thermo Scientific Pierce TPCK Trypsin is a serine endoprotease that is applicable to amino acid analysis and protein sequencing, mapping and structural studies; the immobilized form allows sample separation after treatment.
- Cleaves at carboxyl side of arginine and lysine residues
- Digestion conditions: pH 7.5-9.0, 37°
Immobilized TPCK Trypsin can be substituted for free trypsin in many application and is advantageous because it minimizes autolysis, eliminates contamination of a sample with the protease and allows control of the digestion by removing the trypsin. Immobilized trypsin is also more stable against heat-induced denaturation, resulting in longer maintenance of activity.
Ordering information: Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8° C.
Caution: For Research Use Only. Not for use in diagnostic procedures.