Order Entry
Puerto Rico
ContactUsLinkComponent
Trypsin, Agarose
Trypsin, Agarose
Catalog # PI20230
Supplier:  Invitrogen
CAS Number:  
undefined
Trypsin, Agarose
Catalog # PI20230
Supplier:  Invitrogen
Supplier Number:  20230
CAS Number:  

Specifications

  • Conjugation:
    Agarose
  • Size:
    2 ml
  • Storage Conditions:
    Upon receipt store at 4 ˚C.
  • Shipping Conditions:
    ambient
  • Enzyme Name:
    Trypsin
  • Cat. No.:
    PI20230
  • Supplier no.:
    20230

Specifications

About this item

Thermo Scientific Pierce Immobilized TPCK Trypsin is a serine endoprotease that is applicable to amino acid analysis and protein sequencing, mapping and structural studies; the immobilized form allows sample separation after treatment.

  • Cleaves at carboxyl side of arginine and lysine residues
  • Digestion conditions: pH 7.5-9.0, 37˚
  • Immobilized for ease of separation from cleavage products or affinity purification of Trypsin Inhibitors

Trypsin has a wide range of applications including amino acid analysis and protein sequencing, mapping and structural studies. Enzymes such as trypsin and chymotrypsin have become important tools in sequencing studies since they are highly selective in their cleavage of peptide bonds. Trypsin cleaves only those peptide bonds in which the carboxyl group is contributed by a lysine or an arginine residue, regardless of the length or amino acid sequence of the chain. Our Immobilized TPCK Trypsin can be substituted for free trypsin in many application and is advantageous because it minimizes autolysis, eliminates contamination of a sample with the protease and allows control of the digestion by removing the trypsin. Immobilized trypsin is also more stable against heat-induced denaturation, resulting in longer maintenance of activity.

Caution: For Research Use Only. Not for use in diagnostic procedures.