Pierce™ Streptavidin Coated High Sensitivity Plates, 8-Well Strip, Thermo Scientific
Thermo Scientific Pierce Streptavidin Coated High Sensitivity Plates maximize capture efficiency of Biotinylated probes and control background in assays for low levels of target analyte.
- Sensitive detection of low concentrations (ng/mL) of biotinylated IgG
- Ready to use and pre-blocked with Blocker™ BSA buffer
- Detection range is 5 to 300ng/mL (100μL) of biotinylated IgG
- Activation level is 250μL
Streptavidin is the most popular and widely available biotin-binding protein for research methods
Clear plates for colorimetric ELISA and other assays. Black plates for fluorescent assays.
Streptavidin has an isoelectric point of 5 to 6, resulting in low non-specific interactions.
: Upon receipt store at 4 °C. Product is shipped at ambient temperature.
: For Research Use Only. Not for use in diagnostic procedures.