Duck IgY Isotype Control
10013-950
- Antibody Type:Control
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Duck
- Isotype:IgY
- Reactivity:Duck
- Size:5 mg
- Format:Purified IgG
- Form:Liquid
- Storage Buffer:10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage Temperature:2 to 8°C
- Concentration:8-12 mg/mL
- Shipping Temperature:2 to 8°C
- Purification:Affinity Purified
- Cat. No.:10013-950
- Supplier no.:IR2896
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.
- Host: Duck
- Conjugation: Unconjugated
- Clonality: Polyclonal
- Isotype: IgY
- Reactivity: Duck IgY
- Concentration: 8-12 mg/mL
- Purification Method: Affinity Purified
- Form: Liquid
- Storage Buffer: 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage Temperature: 2 to 8°C
- Shipping Temperature: 2 to 8°C
- Format: Purified IgG
- ELISA: Yes
- Western Blotting: Yes
This product is used as a blocking reagent or control for most immunoassay applications.