Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Lethal Factor from Bacillus anthracis Chicken Polyclonal Antibody (Biotin)
Anti-Lethal Factor from Bacillus anthracis Chicken Polyclonal Antibody (Biotin)
Catalog # 102946-628
Anti-Lethal Factor from Bacillus anthracis Chicken Polyclonal Antibody (Biotin)
Catalog # 102946-628
Supplier Number:  768L

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Lethal Factor from Bacillus anthracis
  • Antigen Symbol:
    Lethal Factor from Bacillus anthracis
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Host:
    Chicken
  • Isotype:
    lgY
  • Reactivity:
    Chicken
  • Size:
    100 µg
  • Storage Temperature:
    2°C - 8°C
  • Concentration:
    1 mg/ml
  • Shipping Temperature:
    Ships on blue ice, overnight.
  • Immunogen:
    LF from B. anthracis
  • Cat. No.:
    102946-628
  • Supplier no.:
    768L

Specifications

About this item