Human Recombinant UBCH10
Catalog # 10033-646
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:100 µg
- Protein/Peptide Name:UBCH10
- Purity:>95% (SDS-PAGE)
- Molecular Weight:~19.7kDa
- Concentration:0.5mg/ml
- Formulation:Liquid. In PBS.
- Cat. No.:10033-646
- Supplier no.:UW0960-0100
Specifications
About this item
Produced in E. coli.