Hydroxytyrosol Acetate, Enzo Life Sciences
Catalog # 89149-704
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- CAS No.:69039-02-7
- Description:Hydroxytyrosol acetate (antioxidant), C10H12O4
- Size:50 mg
- Purity:≥98% (HPLC)
- Molecular Weight:196.2
- Cat. No.:89149-704
- Supplier no.:350-404-M050
Specifications
About this item
Antioxidant
Synthetic
Antioxidant comparable to resveratrol (Prod. No. BML-FR104). Is converted to hydroxytyrosol through hydrolysis by esterases. Reaches full oxygen radical absorbance capacity (ORAC) when converted to free hydoxytyrosol. ORAC values of hydroxytyrosol are about 50% higher than resveratrol, 10-fold higher than L(+)-ascorbic acid and 4-fold higher than (-)-epicatechin. Has been shown to have anticancer and antiviral effects.Antibacterial at 400µg/mL against Streptococcus pyogenes, Staphylococcus aureus, Escherichia coli and Klebsiella pneumoniae.