Orders LinkContactUsLinkComponent
RDO Rapid Decalcifier, Electron Microscopy Sciences
RDO Rapid Decalcifier, Electron Microscopy Sciences
Certificates
RDO Rapid Decalcifier, Electron Microscopy Sciences
RDO Rapid Decalcifier, Electron Microscopy Sciences
8 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
RDO and RDO Gold were developed with the basic premise of making the work routine quicker and easier without sacraficing the quality and integrity of samples. These are remarkably effective multipurpose decacifiers that can be tailored to suit specific lab routine.
  • Safe, non toxic and non-corrosive
  • Dilutable and translucent
  • Microwavable and biodegradable
  • Great cellular differentiation
Both RDO and RDO gold provides crisp nuclear staining and definition, give distinguishable chromatin, show great surface decalcification, safe enough to be disposed down to lab sink to drain.
RDO Gold is designated for biopsies which contain minimal amounts of calcium. It requires no filtration and gives stellar morphological representation of bone biopsies. There is no folding of tissues with RDO Gold and it is clear and non-precipitate.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 102094-938
    RDO Decalcifier
    RDO Decalcifier
    1 l
    Log in for availability
    Log in for availability
    Each
    Each - $53.32
    $53.32
    Each
       $53.32
    Supplier Number: 64143-01
  • 102094-940
    RDO Decalcifier
    RDO Decalcifier
    6×1 l
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $213.13
    $213.13
    Case of 6
       $213.13
    Supplier Number: 64143-06
  • 102094-942
    RDO Decalcifier
    RDO Decalcifier
    1 gal.
    Log in for availability
    Log in for availability
    Each
    Each - $133.28
    $133.28
    Each
       $133.28
    Supplier Number: 64143-40
  • 102094-944
    RDO Decalcifier
    RDO Decalcifier
    4×1 gal.
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $420.60
    $420.60
    Case of 4
       $420.60
    Supplier Number: 64143-44
  • 102094-946
    RDO-Gold Decalcifier
    RDO-Gold Decalcifier
    1 l
    Log in for availability
    Log in for availability
    Each
    Each - $70.42
    $70.42
    Each
       $70.42
    Supplier Number: 64144-01
  • 102094-948
    RDO-Gold Decalcifier
    RDO-Gold Decalcifier
    6×1 l
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $368.02
    $368.02
    Case of 6
       $368.02
    Supplier Number: 64144-06
  • 102094-950
    RDO-Gold Decalcifier
    RDO-Gold Decalcifier
    1 gal.
    Log in for availability
    Log in for availability
    Each
    Each - $176.80
    $176.80
    Each
       $176.80
    Supplier Number: 64144-40
  • 102094-952
    RDO-Gold Decalcifier
    RDO-Gold Decalcifier
    4×1 gal.
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $552.87
    $552.87
    Case of 4
       $552.87
    Supplier Number: 64144-44