Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium iodide 3% in aqueous solution
Health
Potassium iodide 3% in aqueous solution
Catalog # 101410-850
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Health
Potassium iodide 3% in aqueous solution
Catalog # 101410-850
Supplier Number:  25109-3
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    500 ml
  • Cat. No.:
    101410-850
  • Supplier no.:
    25109-3

Specifications

About this item

 :
 :166 g/mol
Melting Pt:681 °C (1258 °F)
 :GHS08
Storage Temperature:Ambient
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405