Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Haematoxylin solution (Heidenhain)

Haematoxylin solution (Heidenhain)

Bottle, 500 ml
Catalog # 101410-914
Supplier #:  26042-20
Haematoxylin solution (Heidenhain)
Catalog # 101410-914
CAS Number:  517-28-2
Haematoxylin solution (Heidenhain)
Haematoxylin solution (Heidenhain)

Bottle, 500 ml

CAS Number:  517-28-2
Price
$52.07
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Hematoxylin solution is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the biological staining commission and the armed forces institute of pathology.

  • Available in concentrations of 1.0% aqueous, 5% in SD alcohol, in 95% alcohol, and as Heidenhain's hematoxylin
Synonyms:  DcH-48, Natural Black I, Hydroxybrasilin
  • CAS Number:517-28-2
  • Formula:C₁₆H₁₄O₆
  • MDL Number:MFCD00078111
  • Molecular Weight:302.28 g/mol
  • Boiling Pt:100 °C (212 °F)
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.