Herxheimer's Technique, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Herxheimer’s Technique is used to stain fat and cholesterol
Stain results - Nuclei: Blue, Fat: Orange-Red, Cholesterol: Red.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.