Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Nuclear Fast Red solution

Nuclear Fast Red solution

500 ml
Catalog # 100504-576
Supplier #:  26212-03
Nuclear Fast Red solution
Catalog # 100504-576
CAS Number:  6409-77-4
Nuclear Fast Red solution
Nuclear Fast Red solution

500 ml

CAS Number:  6409-77-4
Price
$95.48
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Pizzolato’s Method for Calcium Oxalate is used for bone and its related stain.
Stain results - Calcium oxalate: Black, All other tissue elements: Red.
Solutions to use, Silver Nitrate 5% Aqueous, Hydrogen Peroxide 30%, Nuclear Fast Red (Kernechtrot) Solution
Pizzolato, P., J. Histochem. Cytochem., 12:333-136, 1964.
Luna, L.G., (ed), Manual of Staining Methods of the AFIP, 3rd Ed, McGraw-Hill, NY, c 1968, p 77.

Synonyms:  CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
  • CAS Number:6409-77-4
  • Formula:C₁₄H₈NNaO₇S
  • MDL Number:MFCD00078493
  • Molecular Weight:357.28 g/mol
Specifications

Specifications

  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.