Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine dihydrochloride ≥98%

3,3',5,5'-Tetramethylbenzidine dihydrochloride ≥98%

Irritant
Health
3,3',5,5'-Tetramethylbenzidine dihydrochloride ≥98%
Certificates
3,3',5,5'-Tetramethylbenzidine dihydrochloride ≥98%

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

C16H20N2·2HCl F.W. 313.30 Assay 98% (Cl) Also available in tablet form. 1mg/ tablet, ready to use.

  • TMB-HCl is also available in 1 mg tablets that are ready to use
Synonyms:  Dihydrochloride TMB, TMB-HCl, TMB 2HCl, TMB dihydrochloride
  • CAS Number:64285-73-0
  • Formula:C₁₆H₂₀N₂·2HCl
  • MDL Number:MFCD00012961
  • Molecular Weight:313.27 g/mol
  • Melting Pt:300 °C (572 °F)
  • Storage Temperature:Refrigerator
Specifications for Product Family

Specification Test Results

  • Assay:
    98% (Cl)
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    10 Tablets
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $91.62
    $91.62
    Pack of 10
       $91.62

    Supplier Number: 13075
  • Bottle
    50 Tablets
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $474.38
    $474.38
    Pack of 50
       $474.38

    Supplier Number: 13076
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g/mol)
    Each (1g/mol) - $192.10
    $192.10
    Each (1g/mol)
       $192.10

    Supplier Number: 13074

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.