About this item
Hatmann’s (Davidson’s) fixative is rapid fixative. Results with good nuclear detail and minimal formalin pigment.
This is rapid fixative. Results with good nuclear detail and minimal formalin pigment. Hatmann’s (Davidson’s) Fixative is useful for preparing tumors, bone marrow, gynecologic specimen, fatty breast, medical biopsies. Fixation of small specimen is rapid. Exposure to the fixative should be limited to 24 hours (tissue may then be transferred to neutral buffered formalin or 70% alcohol for storage).
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- UN No.:1986
- Storage Temperature:RT
- Density:0.9501 g/cm³ (7.929 lbs/gal)
- Flash Point:17 °C (63 °F)
- Boiling Point:-21 °C (-6 °F)