Order Entry
Puerto Rico
ContactUsLinkComponent
Hydrochloric acid solution 0.1 N
Corrosive
Hydrochloric acid solution 0.1 N
Catalog # 100505-060
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Corrosive
Hydrochloric acid solution 0.1 N
Catalog # 100505-060
Supplier Number:  16760
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    100 ml
  • Cat. No.:
    100505-060
  • Supplier no.:
    16760

Specifications

About this item

 :
Boiling Pt:100 °C (212 °F)
Density:1.003 g/cm³ (8.37 lbs/gal.)
Storage Temperature:Ambient
ADR:8,III
 :36.46
UN:1789
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324