Order Entry
Puerto Rico
ContactUsLinkComponent
Hematoxylin, certified (C.I.75290, DcH-48)
Toxic
Irritant
Hematoxylin, certified (C.I.75290, DcH-48)
Catalog # 100505-024
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Toxic
Irritant
Hematoxylin, certified (C.I.75290, DcH-48)
Catalog # 100505-024
Supplier Number:  16620
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    10 g
  • Cat. No.:
    100505-024
  • Supplier no.:
    16620

Specifications

About this item

 :
 :302.28 g/mol
 :GHS06
 :GHS07
Storage Temperature:Ambient
CAS Number:517-28-2
 :208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111