Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Formaldehyde 37% in aqueous solution, microfiltered

10×10 ml
  100503-912
 
 
 
 
 
Formaldehyde 37% in aqueous solution, microfiltered
  100503-912
 :  50-00-0
Formaldehyde 37% in aqueous solution, microfiltered
Formaldehyde 37% in aqueous solution, microfiltered

10×10 ml

 :  50-00-0
 
$71.31
   
Each (10ml)
 
   
 

Microfiltered with a 0.45 micron membrane and suitable as an EM fixative when a methanol free solution is not necessary.

Available in 10%, 16%, 20%, and 32% solutions.

 :  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  •  :50-00-0
  •  :CH₂O
  •  :MFCD00003274
  •  :30.03 g/mol
  •  :64 °C (147 °F)
  •  :0.9045 g/cm³ (7.548 lbs/gal.)
  •  :60 °C (140 °F
  •  :Ambient
  •  :1198
  •  :3,III
 

 

  • Size:
    10×10 ml

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.