About this item
All of our Earle’s Balanced Salt Solution contains: Potassium Chloride, Sodium Chloride, Sodium Phosphate Monobasic, and D-glucose. Some formulations may contain additional components. All of the solutions are prepared 1X strength.
Earle’s Balanced Salt Solutions Formula I contains:
KCl: 0.157 M
NaCl: 0.157 M
NaH₂PO₄·H₂O: 0.157 M
MgSO₄·7H₂O: 0.157 M
CaCl₂,·2H₂O: 0.157 M
NaHCO₃: 0.157 M
D-Glusose: 1 g/L
Phenol Red: none