Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) ≥98%

3,3',5,5'-Tetramethylbenzidine (TMB) ≥98%

1 g
Catalog # 100503-800
Irritant
Supplier #:  13070
3,3',5,5'-Tetramethylbenzidine (TMB) ≥98%
Catalog # 100503-800
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) ≥98%
3,3',5,5'-Tetramethylbenzidine (TMB) ≥98%

1 g

CAS Number:  54827-17-7
Price
$79.07
Log in for availability
Each (1g/mol)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

A non-carcinogenic substitute for DAB and DAB-HCl.

Can be used as a sensitive and specific reagent for the detection of blood (Standefer, J.C., Vanderjagt, D. ,1977; Assay of Hemoglobin. Clin. Chem. 23, 749; Boss, E.S., et al., (1981); Assay of Peroxidases. J. of Immunoassay. 2,187; Holland, V.R., et al., Tetrahedron, 30, 3299, 1974).

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specification Test Results

  • Assay:
    98%

Specifications

  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.