Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Agarose S FR White, Fine Powder
- Size:100 g
- Cat. No.:100503-644
- Supplier no.:10207
- CAS No.:9012-36-6
Specifications
About this item
(C6H10O5·C6H8O4)n CAS 9012-36-6
Agarose SFR™ is a high resolution sieving agarose with unsurpassed clarity. DNA bands differing in size by 2% can be resolved in the range of 200 – 1,000 bp. This agarose is suitable for the analysis of AFLP’s (Amplified Fragment Length Polymorphisms), STR’s (Short Tandem Repeats) and tetranucleotide repeats. The low melting point of agarose SFR™ makes it an excellent medium for analytical and preparative electrophoresis. ™ trademark of Amresco®