Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Acetone ACS

Flammable
Irritant
Acetone ACS
Certificates
Acetone ACS

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CH3COCH3 F.W. 58.08 CAS 67-64-1 A.C.S.

Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Boiling Pt:56.5 °C (134 °F)
  • Melting Pt:94.7 °C (202 °F)
  • Density:0.79 g/cm³
  • Flash Pt:–20 °C (–4 °F)
  • UN:1090
  • ADR:3,II
Specifications for Product Family

Specification Test Results

  • Aldehyde (as HCHO):
    0.002%
  • Color:
    Max. APHA 10
  • Isopropyl Alchohol:
    0.05%
  • Methanol (CH₃OH):
    0.05%
  • Residue after Evaporation:
    0.001%
  • Titratable Acid:
    0.0003 meq/g
  • Titratable Base:
    0.0006 meq/g
  • Water:
    0.5%
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    250 ml
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $21.07
    $21.07
    Each (250ml)
       $21.07

    Supplier Number: 10012
  • Bottle
    4×250 ml
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $76.72
    $76.72
    Each (250ml)
       $76.72

    Supplier Number: 10014
  • Bottle
    946 ml
    Log in for availability
    Log in for availability
    Each
    Each - $35.02
    $35.02
    Each
       $35.02

    Supplier Number: 10000
  • Bottle
    4×946 ml
    Log in for availability
    Log in for availability
    Each (1qt US)
    Each (1qt US) - $128.63
    $128.63
    Each (1qt US)
       $128.63

    Supplier Number: 10010

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.