
1,4-Butanediol diglycidyl ether
Catalog # 100503-728
Supplier: Electron Microscopy Sciences
Minority or Woman-Owned Business Enterprise
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Bottle
- Size:100 ml
- Cat. No.:100503-728
- Supplier no.:11800
Specifications
Specification Test Results
- Specific Gravity:1.10
Specification Test Results
About this item
:
:202.25 g/mol
Boiling Pt:266 °C (511 °F)
Density:1.05 g/cm³ (8.762 lbs/gal.)
Flash Pt:>–112 °C (>–234 °F)
:GHS07
CAS Number:2425-79-8
Formula:C₁₀H₁₈O₄
MDL Number:MFCD00005146