ContactUsLinkComponent
Sklar Black™ Adson Dressing Forceps, OR Grade, Sklar
Sklar Black™ Adson Dressing Forceps, OR Grade, Sklar
Catalog # 10199-092
Supplier:  Sklar
undefined
Sklar Black™ Adson Dressing Forceps, OR Grade, Sklar
Catalog # 10199-092
Supplier:  Sklar
Supplier Number:  90-2261
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    121 mm (4¾")
  • Material:
    Stainless Steel with Reduced Reflectivity Polymer Coating
  • Tip configuration:
    2 x 3 Teeth
  • Cat. No.:
    10199-092
  • Jaw type:
    Straight
  • Tip style:
    Serrated
  • Supplier no.:
    90-2261
Specifications
About this item
Sklar Black™ Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.
They have a wide thumb grasp for increased precision and control. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed for laser surgeries.
This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
Certifications: Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects.