ContactUsLinkComponent
Bandage Scissors, OR Grade, Sklar
Bandage Scissors, OR Grade, Sklar
  89025-446
 :  Sklar
undefined
Bandage Scissors, OR Grade, Sklar
  89025-446
 :  Sklar
 :  11-1252
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Hi-Level Bandage Scissors, Premium OR-Grade
  • Length:
    13.9 cm (5.5")
  • Material:
    Stainless steel
  • Curvature:
    Straight Blades with Angled Shanks
  • Cat. No.:
    89025-446
  • Sterility:
    Non sterile
  • Supplier no.:
    11-1252
  • Blade type:
    Sharp/Blunt, Serrated
Sklar's® Hi-Level Bandage Scissors are primarily used for removing tight bandages.
The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin
Censitrac Ready products are industry standard and constructed of stainless steel.
This product has one sharp tip along with the nodule tip. The shanks are strongly curved before the screw lock and the blades are straight.
 : Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
 : Customers must have a Censitrac Site ID to purchase Censitrac instruments.