Specifications
- Length:89 mm (3½")
- Material:Stainless steel
- Cat. No.:10194-540
- Supplier no.:23-1196
- Blade type:Straight, Smooth, Blunt/Blunt
Specifications
About this item
Sklarlite XD™ Shortbent Suture scissors are a lighter, more delicate version of standard Shortbent scissors used for suture removal.
A unique manufacturing process developed by shaves microns off standard and delicate pattern instruments, while maintaining structural integrity and durability.
This lightweight modification provides the surgeon with the highest degree of precision with the least amount of fatigue.