Order Entry
Puerto Rico
ContactUsLinkComponent
Tischler-Kevorkian Micro-Cut Biopsy Punch, OR Grade, Sklar
Tischler-Kevorkian Micro-Cut Biopsy Punch, OR Grade, Sklar
Catalog # 10194-148
Supplier:  Sklar
undefined
Tischler-Kevorkian Micro-Cut Biopsy Punch, OR Grade, Sklar
Catalog # 10194-148
Supplier:  Sklar
Supplier Number:  90-8920

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Biopsy punch, rectangular, straight blade, rectangular bite with 1 lower tooth
  • Length:
    203 mm (8")
  • Material:
    Stainless steel
  • Cat. No.:
    10194-148
  • Supplier no.:
    90-8920

Specifications

About this item

Tischler-Kevorkian Micro-Cut Biopsy Punch is an OB/GYN instrument used in colposcopy procedures to obtain cervical tissue samples for biopsy.

This product features a single lower tooth and a single upper tooth used to grasp difficult to reach tissue. It also has a locking mechanism above the horn on the handle, so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal.

This reduces the risk of losing the tissue sample as the instrument is withdrawn. This product has a pistol style handle with one large ring for multiple fingers.