Anti-AOC3 Mouse Polyclonal Antibody
Catalog # 10559-010
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Amine Oxidase, Copper Containing 3
- Antigen Symbol:AOC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:VAP-1.|Vascular adhesion protein 1|Semicarbazide-sensitive amine oxidase|Membrane primary amine oxidase|Membrane copper amine oxidase|Semicarbazide sensitive amine oxidase|Vascular Adhesion Protein 1|HPAO|SSAO|AOC3|VP97|AOC3_HUMAN|Amine Oxidase|Copper Containing 3 (vascular adhesion protein 1)|Copper amine oxidase
- Storage Buffer:No additive
- Storage Temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:AOC3 (AAH50549.1, 1 a.a. ~ 763 a.a) full-length human protein.
- Cat. No.:10559-010
- Supplier no.:H00008639-B01
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human AOC3 protein.
Recommended Dilutons: Western Blot (1:500-1:1000)
Type: Primary
Antigen: AOC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human