Anti-DYRK2 Mouse Monoclonal Antibody [clone: 2F9]
10624-212
: Abnova
- Antibody Type:Primary
- Antigen Name:Dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2
- Antigen Symbol:DYRK2
- Clonality:Monoclonal
- Clone:2F9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:200 µL
- Cross Adsorption:No
- Form:Liquid
- Storage Buffer:In ascites fluid
- Sequence:MNDHLHVGSHAHGQIQVQQLFEDNSNKRTVLTTQPNGLTTVGKTGLPVVPERQLDSIHRRQGSSTSLKSMEGMGKVKATPMTPEQAMKQYMQKLTAFEHH
- Storage Temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:DYRK2 (AAH05809, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Cat. No.:10624-212
- Supplier no.:H00008445-M04A
Mouse monoclonal antibody raised against a partial recombinant DYRK2.
Type: Primary
Antigen: DYRK2
Clonality: Monoclonal
Clone: 2F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human