Anti-AOC2 Rabbit Polyclonal Antibody
Catalog # 10577-018
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Amine Oxidase, Copper Containing 2
- Antigen Symbol:AOC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:copper containing 2 retina-specific|RAO|DAO2|amine oxidase
- Storage Buffer:No additive
- Storage Temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:AOC2 (NP_033720.2, 1 a.a. ~ 756 a.a) full-length human protein.
- Cat. No.:10577-018
- Supplier no.:H00000314-D01
Specifications
About this item
Rabbit polyclonal antibody raised against a full-length human AOC2 protein.
Recommended Dilutons: Western Blot (1:1000)
Type: Primary
Antigen: AOC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human