Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-AOC2 Mouse Polyclonal Antibody
Anti-AOC2 Mouse Polyclonal Antibody
Catalog # 10559-008
Supplier:  Abnova
undefined
Anti-AOC2 Mouse Polyclonal Antibody
Catalog # 10559-008
Supplier:  Abnova
Supplier Number:  H00000314-B01P

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Amine Oxidase, Copper Containing 2
  • Antigen Symbol:
    AOC2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 µg
  • Cross Adsorption:
    No
  • Form:
    Liquid
  • Antigen Synonyms:
    copper containing 2 retina-specific|RAO|DAO2|amine oxidase
  • Storage Buffer:
    In 1x PBS, pH 7.4
  • Storage Temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    AOC2 (NP_033720.2, 1 a.a. ~ 756 a.a) full-length human protein.
  • Cat. No.:
    10559-008
  • Supplier no.:
    H00000314-B01P

Specifications

About this item

Mouse polyclonal antibody raised against a full-length human AOC2 protein.

Recommended Dilutons: Western Blot (1 ug/mL)

Type: Primary
Antigen: AOC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human