Order Entry
Puerto Rico
ContactUsLinkComponent
Innovera® E285A Toner Cartridge, Essendant
Innovera® E285A Toner Cartridge, Essendant
Catalog # 102407-734
undefined
Innovera® E285A Toner Cartridge, Essendant
Catalog # 102407-734
Supplier Number:  IVRE285A

Specifications

  • Cartridge no.:
    85A
  • For Use With:
    HP LaserJet Pro M1130, M1132MFP, M1134MFP, M1136MFP, M1137, M1138, M1139, M1210, M1212NF, M1212F, M1213NF, M1214NFH, M1216NFH, M1217NFW, M1219NF, P1100, P1102, P1102W
  • Yield:
    1600 pages
  • Color:
    Black
  • Cat. No.:
    102407-734
  • Supplier no.:
    IVRE285A

Specifications

About this item

Guarantees great prints with low cost.

  • Consistent and reliable performance
  • Compatible and remanufactured
  • Easy to install, reliable and dependable

Manufactured and tested to provide the same quality, performance and page yield.