Order Entry
Puerto Rico
ContactUsLinkComponent
Boardwalk® JRT Jr. Bathroom Tissue
Boardwalk® JRT Jr. Bathroom Tissue
Catalog # 102866-162
undefined
Boardwalk® JRT Jr. Bathroom Tissue
Catalog # 102866-162
Supplier Number:  BWK6101

Specifications

  • Ø:
    9.125"
  • Width:
    3.625"
  • Length:
    2000'
  • Cat. No.:
    102866-162
  • Supplier no.:
    BWK6101
  • Ply:
    1

Specifications

About this item

Economical roll is produced using recycled fibers.

  • Embossed for added softness and absorbency