- Description:Little Retractor with Extra Thin Blade
- Length:18.4 cm (7¼")
- Material:Stainless steel
- Cat. No.:10197-776
- Supplier no.:60-5472
Sklar's® Little Retractor is a single-end, handheld instrument used to hold back intermediate depth tissue
The ½" wide blade is smooth with a rounded tip and strong curve. The Little Retractor features a flat, grooved handle.