Order Entry
Puerto Rico
ContactUsLinkComponent
 
Innovera® H562WN Ink Cartridge, Essendant
  102407-798
undefined
Innovera® H562WN Ink Cartridge, Essendant
  102407-798
 :  IVRH562WN

 

  • Cartridge no.:
    61
  • For Use With:
    HP Deskjet 1000, 1050, 1055, 2050, 3000, 3050, 3050A, 3052A, 3054A
  • Yield:
    165 pages
  • Color:
    Tri-Color
  • Cat. No.:
    102407-798
  • Supplier no.:
    IVRH562WN

 

 

Guarantees great prints with low cost.

  • Compatible and remanufactured
  • Easy to install, reliable and dependable

Manufactured and tested to provide the same quality, performance and page yield.