Order Entry
Puerto Rico
ContactUsLinkComponent
Stitch Scissors, OR Grade, Sklar
Stitch Scissors, OR Grade, Sklar
Catalog # 89025-340
Supplier:  Sklar
undefined
Stitch Scissors, OR Grade, Sklar
Catalog # 89025-340
Supplier:  Sklar
Supplier Number:  22-2950

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Length:
    127 mm (5")
  • Material:
    Stainless steel
  • Cat. No.:
    89025-340
  • Supplier no.:
    22-2950
  • Blade type:
    Curved, Blunt/Blunt

Specifications

About this item

Stitch Scissors are used for suture removal.

These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting.

Stitch Scissors should not be used for cutting tissue.